3-(Ethoxymethyl)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023497-500MG
101816-122EA 235.75 USD
101816-122
3-(Ethoxymethyl)pyrrolidine hydrochloride
3-(Ethoxymethyl)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNO
MDL Number: MFCD09997927
CAS Number: 1185298-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR