3-[(3-Phenylpropoxy)methyl]pyrrolidine

Supplier: Matrix Scientific

023496-500MG
101816-120EA 315.38 USD
101816-120
3-[(3-Phenylpropoxy)methyl]pyrrolidine
3-[(3-Phenylpropoxy)methyl]pyrrolidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD08688481
CAS Number: 946682-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR