3-(Propoxymethyl)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023495-500MG
101816-118EA 235.75 USD
101816-118
3-(Propoxymethyl)pyrrolidine hydrochloride
3-(Propoxymethyl)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₈H₁₈ClNO
MDL Number: MFCD12405059
CAS Number: 946716-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR