4-[2-(1-Naphthyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

023493-500MG
101816-114EA 315.38 USD
101816-114
4-[2-(1-Naphthyloxy)ethyl]piperidine hydrochloride
4-[2-(1-Naphthyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₂ClNO
MDL Number: MFCD22580726
CAS Number: 1417567-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR