4-[2-(2,3-Dihydro-1H-inden-5-yloxy)ethyl]piperidine

Supplier: Matrix Scientific

023492-500MG
101816-112EA 315.38 USD
101816-112
4-[2-(2,3-Dihydro-1H-inden-5-yloxy)ethyl]piperidine
4-[2-(2,3-Dihydro-1H-inden-5-yloxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₆H₂₃NO
MW: 245.37 g/mol
MDL Number: MFCD08687586
CAS Number: 946682-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR