4-{2-[4-(tert-Butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

023490-500MG
101816-108EA 235.75 USD
101816-108
4-{2-[4-(tert-Butyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[4-(tert-Butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD11100512
CAS Number: 1185299-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR