4-[2-(3-Methylphenoxy)ethyl]piperidine

Supplier: Matrix Scientific

023487-500MG
101816-102EA 315.38 USD
101816-102
4-[2-(3-Methylphenoxy)ethyl]piperidine
4-[2-(3-Methylphenoxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD08688325
CAS Number: 946682-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR