4-[2-(Benzyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

023482-500MG
101816-092EA 370.3 USD
101816-092
4-[2-(Benzyloxy)ethyl]piperidine hydrochloride
4-[2-(Benzyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD09879250
CAS Number: 906744-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR