4-[2-(3-Phenylpropoxy)ethyl]piperidine

Supplier: Matrix Scientific

023479-500MG
101816-086EA 235.75 USD
101816-086
4-[2-(3-Phenylpropoxy)ethyl]piperidine
4-[2-(3-Phenylpropoxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₆H₂₅NO
MDL Number: MFCD08688480
CAS Number: 907159-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR