3-[2-(1-Naphthyloxy)ethyl]piperidine

Supplier: Matrix Scientific

023476-500MG
101816-080EA 315.38 USD
101816-080
3-[2-(1-Naphthyloxy)ethyl]piperidine
3-[2-(1-Naphthyloxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₇H₂₁NO
MDL Number: MFCD08688425
CAS Number: 946682-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR