4-Methoxyphenyl-2-(3-piperidinyl)ethyl ether

Supplier: Matrix Scientific

023472-500MG
101816-072EA 235.75 USD
101816-072
4-Methoxyphenyl-2-(3-piperidinyl)ethyl ether
4-Methoxyphenyl-2-(3-piperidinyl)ethyl ether

Hazard: Irritant

Formula: C₁₄H₂₁NO₂
MDL Number: MFCD08688102
CAS Number: 946682-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR