3-[2-(3-Fluorophenoxy)ethyl]piperidine

Supplier: Matrix Scientific

023468-500MG
101816-064EA 315.38 USD
101816-064
3-[2-(3-Fluorophenoxy)ethyl]piperidine
3-[2-(3-Fluorophenoxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₈FNO
MDL Number: MFCD08687882
CAS Number: 946682-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR