3-[2-(Tetrahydro-2-furanylmethoxy)ethyl]piperidine

Supplier: Matrix Scientific

023466-500MG
101816-060EA 315.38 USD
101816-060
3-[2-(Tetrahydro-2-furanylmethoxy)ethyl]piperidine
3-[2-(Tetrahydro-2-furanylmethoxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₂H₂₃NO₂
MDL Number: MFCD08688538
CAS Number: 946760-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR