3-[2-(Benzyloxy)ethyl]piperidine

Supplier: Matrix Scientific

023465-500MG
101816-058EA 315.38 USD
101816-058
3-[2-(Benzyloxy)ethyl]piperidine
3-[2-(Benzyloxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD08686863
CAS Number: 946727-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR