3-[2-(3-Phenylpropoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

023462-500MG
101816-052EA 235.75 USD
101816-052
3-[2-(3-Phenylpropoxy)ethyl]piperidine hydrochloride
3-[2-(3-Phenylpropoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD12026507
CAS Number: 1185298-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR