2-[2-(1-Naphthyloxy)ethyl]piperidine

Supplier: Matrix Scientific

023459-500MG
101816-046EA 315.38 USD
101816-046
2-[2-(1-Naphthyloxy)ethyl]piperidine
2-[2-(1-Naphthyloxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₇H₂₁NO
MDL Number: MFCD08688423
CAS Number: 93723-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR