2-[2-(2,3-Dihydro-1H-inden-5-yloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

023458-500MG
101816-044EA 235.75 USD
101816-044
2-[2-(2,3-Dihydro-1H-inden-5-yloxy)ethyl]piperidine hydrochloride
2-[2-(2,3-Dihydro-1H-inden-5-yloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO
MDL Number: MFCD10687228
CAS Number: 1185303-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR