2-[2-(4-Methylphenoxy)ethyl]piperidine

Supplier: Matrix Scientific

023457-500MG
101816-042EA 315.38 USD
101816-042
2-[2-(4-Methylphenoxy)ethyl]piperidine
2-[2-(4-Methylphenoxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD08688322
CAS Number: 946760-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR