2-[2-(3-Fluorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

023451-500MG
101816-030EA 315.38 USD
101816-030
2-[2-(3-Fluorophenoxy)ethyl]piperidine hydrochloride
2-[2-(3-Fluorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClFNO
MDL Number: MFCD12405053
CAS Number: 1185299-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR