3-(4-Chloro-2-fluorophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023442-500MG
101816-012EA 186.15 USD
101816-012
3-(4-Chloro-2-fluorophenoxy)pyrrolidine hydrochloride
3-(4-Chloro-2-fluorophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₁ClFNO
MDL Number: MFCD17168389
CAS Number: 1354543-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR