3-(4-Bromo-2-methylphenoxy)pyrrolidine

Supplier: Matrix Scientific

023441-500MG
101816-010EA 315.38 USD
101816-010
3-(4-Bromo-2-methylphenoxy)pyrrolidine
3-(4-Bromo-2-methylphenoxy)pyrrolidine

Hazard: Irritant

Formula: C₁₁H₁₄BrNO
MDL Number: MFCD08687033
CAS Number: 946715-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR