1-{4-[4-(3-Pyrrolidinyloxy)phenyl]-1-piperazinyl}-1-ethanone

Supplier: Matrix Scientific

023440-500MG
101816-008EA 235.75 USD
101816-008
1-{4-[4-(3-Pyrrolidinyloxy)phenyl]-1-piperazinyl}-1-ethanone
1-{4-[4-(3-Pyrrolidinyloxy)phenyl]-1-piperazinyl}-1-ethanone

Hazard: Irritant

Formula: C₁₆H₂₃N₃O₂
MDL Number: MFCD08688534
CAS Number: 946760-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR