3-(2,5-Difluorophenoxy)pyrrolidine hydrochloride
Supplier: Matrix Scientific
023438-500MG
101816-004EA
133.85
USD
101816-004
3-(2,5-Difluorophenoxy)pyrrolidine hydrochloride
3-(2,5-Difluorophenoxy)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₂ClF₂NO |
MDL Number:
MFCD09879248 CAS Number: 1185298-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...