3-(3-Ethoxyphenoxy)pyrrolidine

Supplier: Matrix Scientific

023433-500MG
101815-994EA 315.38 USD
101815-994
3-(3-Ethoxyphenoxy)pyrrolidine
3-(3-Ethoxyphenoxy)pyrrolidine

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD08687746
CAS Number: 946715-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR