3-(2-Methoxy-5-nitrophenoxy)pyrrolidine
Supplier: Matrix Scientific
023432-500MG
101815-992EA
315.38
USD
101815-992
3-(2-Methoxy-5-nitrophenoxy)pyrrolidine
3-(2-Methoxy-5-nitrophenoxy)pyrrolidine
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₄ |
MDL Number:
MFCD08688064 CAS Number: 1504252-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...