3-(3,4-Difluorophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023430-500MG
101815-988EA 186.15 USD
101815-988
3-(3,4-Difluorophenoxy)pyrrolidine hydrochloride
3-(3,4-Difluorophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClF₂NO
MDL Number: MFCD09997075
CAS Number: 1184994-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR