3-(2,4-Difluorophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023429-500MG
101815-986EA 438.46 USD
101815-986
3-(2,4-Difluorophenoxy)pyrrolidine hydrochloride
3-(2,4-Difluorophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClF₂NO
MDL Number: MFCD28126193
CAS Number: 946715-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR