3-(3-Methyl-4-nitrophenoxy)pyrrolidine

Supplier: Matrix Scientific

023428-500MG
101815-984EA 235.75 USD
101815-984
3-(3-Methyl-4-nitrophenoxy)pyrrolidine
3-(3-Methyl-4-nitrophenoxy)pyrrolidine

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₃
MDL Number: MFCD08688300
CAS Number: 946760-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR