3-(3-Nitrophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023427-500MG
101815-982EA 538.46 USD
101815-982
3-(3-Nitrophenoxy)pyrrolidine hydrochloride
3-(3-Nitrophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₃
MDL Number: MFCD22581086
CAS Number: 1425040-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR