8-(3-Pyrrolidinyloxy)quinoline dihydrochloride

Supplier: Matrix Scientific

023426-500MG
101815-980EA 315.38 USD
101815-980
8-(3-Pyrrolidinyloxy)quinoline dihydrochloride
8-(3-Pyrrolidinyloxy)quinoline dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₆Cl₂N₂O
MDL Number: MFCD09997074
CAS Number: 1184969-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR