2-Methyl-5-(3-pyrrolidinyloxy)pyridine hydrochloride
Supplier: Matrix Scientific
023424-500MG
101815-976EA
370.3
USD
101815-976
2-Methyl-5-(3-pyrrolidinyloxy)pyridine hydrochloride
2-Methyl-5-(3-pyrrolidinyloxy)pyridine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O |
MDL Number:
MFCD09997073 CAS Number: 1185302-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...