3-[(4-Chloro-1-naphthyl)oxy]pyrrolidine

Supplier: Matrix Scientific

023420-500MG
101815-970EA 315.38 USD
101815-970
3-[(4-Chloro-1-naphthyl)oxy]pyrrolidine
3-[(4-Chloro-1-naphthyl)oxy]pyrrolidine

Hazard: Irritant

Formula: C₁₄H₁₄ClNO
MDL Number: MFCD08687415
CAS Number: 946727-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR