3-[(1-Bromo-2-naphthyl)oxy]pyrrolidine

Supplier: Matrix Scientific

023418-500MG
101815-966EA 235.75 USD
101815-966
3-[(1-Bromo-2-naphthyl)oxy]pyrrolidine
3-[(1-Bromo-2-naphthyl)oxy]pyrrolidine

Hazard: Irritant

Formula: C₁₄H₁₄BrNO
MDL Number: MFCD08687046
CAS Number: 946681-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR