3-(4-Ethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023416-500MG
101815-962EA 495.38 USD
101815-962
3-(4-Ethylphenoxy)pyrrolidine hydrochloride
3-(4-Ethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD11100510
CAS Number: 1185088-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR