Methyl-2-[4-(3-pyrrolidinyloxy)phenyl]acetate
Supplier: Matrix Scientific
023415-500MG
101815-960EA
235.75
USD
101815-960
Methyl-2-[4-(3-pyrrolidinyloxy)phenyl]acetate
Methyl-2-[4-(3-pyrrolidinyloxy)phenyl]acetate
Hazard: Irritant
Formula:
C₁₃H₁₇NO₃ |
MDL Number:
MFCD08688393 CAS Number: 946726-97-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...