3-(4-Benzylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023414-500MG
101815-958EA 315.38 USD
101815-958
3-(4-Benzylphenoxy)pyrrolidine hydrochloride
3-(4-Benzylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD12405051
CAS Number: 1185297-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR