3-(4-Methylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023412-500MG
101815-954EA 315.38 USD
101815-954
3-(4-Methylphenoxy)pyrrolidine hydrochloride
3-(4-Methylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO
MDL Number: MFCD09997926
CAS Number: 1185073-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR