3-(4-Isopropylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023410-500MG
101815-950EA 315.38 USD
101815-950
3-(4-Isopropylphenoxy)pyrrolidine hydrochloride
3-(4-Isopropylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD24849137
CAS Number: 1449117-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR