4-(1-Methyl-1-phenylethyl)phenyl-3-pyrrolidinyl ether

Supplier: Matrix Scientific

023408-500MG
101815-946EA 315.38 USD
101815-946
4-(1-Methyl-1-phenylethyl)phenyl-3-pyrrolidinyl ether
4-(1-Methyl-1-phenylethyl)phenyl-3-pyrrolidinyl ether

Hazard: Irritant

Formula: C₁₉H₂₃NO
MDL Number: MFCD08688345
CAS Number: 946760-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR