(4-Chlorophenyl)[4-(3-pyrrolidinyloxy)phenyl]methanone hydrochloride

Supplier: Matrix Scientific

023406-500MG
101815-942EA 495.38 USD
101815-942
(4-Chlorophenyl)[4-(3-pyrrolidinyloxy)phenyl]methanone hydrochloride
(4-Chlorophenyl)[4-(3-pyrrolidinyloxy)phenyl]methanone hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₇Cl₂NO₂
MDL Number: MFCD11100509
CAS Number: 1185303-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR