3-([1,1'-Biphenyl]-4-yloxy)pyrrolidine hydrochloride
Supplier: Matrix Scientific
023405-500MG
101815-940EA
495.38
USD
101815-940
3-([1,1'-Biphenyl]-4-yloxy)pyrrolidine hydrochloride
3-([1,1'-Biphenyl]-4-yloxy)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₆H₁₈ClNO |
MDL Number:
MFCD11100508 CAS Number: 1185008-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...