3-(4-Ethoxyphenoxy)pyrrolidine
Supplier: Matrix Scientific
023404-500MG
101815-938EA
315.38
USD
101815-938
3-(4-Ethoxyphenoxy)pyrrolidine
3-(4-Ethoxyphenoxy)pyrrolidine
Hazard: Irritant
Formula:
C₁₂H₁₇NO₂ |
MDL Number:
MFCD08687747 CAS Number: 946715-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...