3-(4-Methoxyphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023402-500MG
101815-934EA 186.15 USD
101815-934
3-(4-Methoxyphenoxy)pyrrolidine hydrochloride
3-(4-Methoxyphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO₂
MDL Number: MFCD09998960
CAS Number: 23123-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR