3-(4-Phenoxyphenoxy)pyrrolidine hydrochloride
Supplier: Matrix Scientific
023401-500MG
101815-932EA
495.38
USD
101815-932
3-(4-Phenoxyphenoxy)pyrrolidine hydrochloride
3-(4-Phenoxyphenoxy)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₆H₁₈ClNO₂ |
MDL Number:
MFCD09997071 CAS Number: 1185302-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...