3-(4-Chloro-3,5-dimethylphenoxy)pyrrolidine

Supplier: Matrix Scientific

023400-500MG
101815-930EA 235.75 USD
101815-930
3-(4-Chloro-3,5-dimethylphenoxy)pyrrolidine
3-(4-Chloro-3,5-dimethylphenoxy)pyrrolidine

Hazard: Irritant

Formula: C₁₂H₁₆ClNO
MDL Number: MFCD08687286
CAS Number: 946726-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR