3-(4-Chlorophenoxy)pyrrolidine

Supplier: Matrix Scientific

023399-500MG
101815-928EA 235.75 USD
101815-928
3-(4-Chlorophenoxy)pyrrolidine
3-(4-Chlorophenoxy)pyrrolidine

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
MDL Number: MFCD08687447
CAS Number: 28491-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR