3-(3-Ethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023396-500MG
101815-922EA 370.3 USD
101815-922
3-(3-Ethylphenoxy)pyrrolidine hydrochloride
3-(3-Ethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD11100507
CAS Number: 946681-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR