3-(3,5-Dimethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023394-500MG
101815-918EA 402.5 USD
101815-918
3-(3,5-Dimethylphenoxy)pyrrolidine hydrochloride
3-(3,5-Dimethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD12405050
CAS Number: 28491-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR