3-(3-Methylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023392-500MG
101815-914EA 370.3 USD
101815-914
3-(3-Methylphenoxy)pyrrolidine hydrochloride
3-(3-Methylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO
MDL Number: MFCD09879242
CAS Number: 1185051-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR