3-(3-Methoxyphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023388-500MG
101815-906EA 538.46 USD
101815-906
3-(3-Methoxyphenoxy)pyrrolidine hydrochloride
3-(3-Methoxyphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO₂
MDL Number: MFCD09879241
CAS Number: 23123-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR